EMAIL THIS PAGE TO A FRIEND
The Journal of the Association of Physicians of India
Barium sulfide poisoning.
PMID 3711009
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
523437
Barium sulfide, 99.9%
BaS