EMAIL THIS PAGE TO A FRIEND
British journal of industrial medicine
Trinitrotoluene: assessment of occupational absorption during manufacture of explosives.
PMID 3718894
Abstract
Trinitrotoluene (TNT) absorption was assessed in groups of workers at two explosives factories by measuring the urinary concentrations of dinitroaminotoluene (DNAT) metabolites. DNAT was detected in most of the urine samples analysed and for postshift samples the mean (SD) concentration was 9.7 (7.9) mg/l (range 0.1-44 mg/l (n = 219)). Individual workers showed substantial day to day variations in DNAT concentrations in postshift urine samples, but on a group basis the concentrations remained fairly constant throughout the working week. Preshift urine samples taken at the beginning of a working week showed low concentrations of DNAT which initially suggested that the elimination of TNT metabolites is fairly rapid. A survey carried out of preshift and postshift urine samples collected from a group of workers for a full working week showed wide variations in the rate of clearance of TNT metabolites from the body and in some cases higher concentrations of metabolites were seen in the samples taken the morning after exposure. When urine samples were collected from the same group of workers after 17 days away from the workplace DNAT was still detectable in samples from eight of the nine subjects, indicating that a proportion of TNT or its metabolites is slowly excreted. When five subjects were monitored more intensively during two workshifts TNT was shown to be absorbed rapidly during the exposure period. In most cases the highest concentrations were seen in the postshift urine samples but significant proportions were still present in samples taken the morning after exposure. Atmospheric levels of TNT were found to be too low to account for the observed excretion of DNAT and dermal uptake rather than inhalation appears to be the major route of absorption.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ERA-018
4-Amino-2,6-dinitrotoluene, vial of 100 mg, analytical standard
C7H7N3O4