EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Enzyme electrode for the measurement of lactate in undiluted blood.
PMID 3719998
Abstract
An enzyme electrode is described based upon the enzyme lactate oxidase (EC 1.1.3.2) coupled to an H2O2 sensor. Use of an organosilane-treated microporous membrane over the enzyme layer, allows responses linear to 18 mmol/l L-lactate with response times of 1-3 min, whilst giving independence from stirring. The method is suitable for the bedside analysis of a single drop of undiluted whole blood. Correlation with a routine method gave y = 0.96x + 0.11, r = 0.996, n = 46.