EMAIL THIS PAGE TO A FRIEND
Nederlands tijdschrift voor geneeskunde
[Hobby and occupation as causes of cancer].
PMID 3724891
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27240
Chrysoidine G, for microscopy (Bact., Bot., Vit.)
C12H12N4 · HCl
80028
Chrysoidine G, analytical standard
C12H12N4 · HCl