EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Artificial reductant enhancement of the Lowry method for protein determination.
PMID 3728976
Abstract
Addition of dithiothreitol in the Lowry procedure 3 min after adding the Folin-Ciocalteau reagent produces immediate color development, with 35 to 60% greater absorbance per mass of protein used.