EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Selective medium for isolation of Fusarium species and dematiaceous hyphomycetes from cereals.
PMID 3729399
Abstract
A selective medium containing 2 micrograms of dichloran per ml, 200 micrograms of chloramphenicol per ml, and 1.5% bacteriological peptone was developed for the isolation of Fusarium species and dematiaceous hyphomycetes from cereals. The medium, designated DCPA, was shown to select against species of Aspergillus, Penicillium, Cladosporium, and mucoraceous fungi. DCPA was evaluated for use as an enumeration medium and compared satisfactorily with dichloran-rose bengal-chloramphenicol agar when both media were tested with a range of cereal samples. Fusarium species and dematiaceous hyphomycetes produced well-formed colonies with good conidial production on DCPA, permitting rapid identification of such isolates on this medium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D67820
2,6-Dichloro-4-nitroaniline, 96%
C6H4Cl2N2O2
45435
Dichloran, PESTANAL®, analytical standard
C6H4Cl2N2O2