EMAIL THIS PAGE TO A FRIEND
Problemy tuberkuleza
[Effect of monoacetylhydrazine on the rat liver].
PMID 3737601
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8309
Acethydrazide, 90%
C2H6N2O