EMAIL THIS PAGE TO A FRIEND
The Analyst
Spectrophotometric determination of bismuth in pharmaceutical preparations using mucic acid.
PMID 3740424
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M89617
Mucic acid, 97%
C6H10O8