EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Streptomyces K15 DD-peptidase-catalysed reactions with suicide beta-lactam carbonyl donors.
PMID 3741377
Abstract
The values of the kinetic parameters that govern the interactions between the Streptomyces K15 DD-peptidase and beta-lactam compounds were determined by measuring the inactivating effect that these compounds exert on the transpeptidase activity of the enzyme and, in the case of [14C]benzylpenicillin and [14C]cefoxitin, by measuring the amounts of acyl-enzyme formed during the reaction. K15 DD-peptidase binds benzylpenicillin or cefoxitin at a molar ratio of 1:1. Benzylpenicilloate is the major product released during breakdown of the acyl-enzyme formed with benzylpenicillin. Benzylpenicillin is not a better acylating agent than the amide Ac2-L-Lys-D-Ala-D-Ala and ester Ac2-L-Lys-D-Ala-D-lactatecarbonyl-donor substrates. beta-Lactam compounds possessing a methoxy group on the alpha-face of the molecule show high inactivating potency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9904
Nα,Nε-Diacetyl-Lys-D-Ala-D-Ala, carboxypeptidase substrate
C16H28N4O6