EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
1,3,5-Trichloro-2-(4-nitrophenoxy)benzene (CNP) in water, sediments, and shellfish of the Ishikari River.
PMID 3742091
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04287
Chlornitrofen, analytical standard
C12H6Cl3N1O3