EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
Acrylonitrile, acrylic and modacrylic fibres, and acrylonitrile-butadiene-styrene and styrene-acrylonitrile copolymers.
PMID 374237