EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Toxicological evaluation of a new nitrification inhibitor and its metabolite].
PMID 3744062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533866
3-Methyl-1-pyrazolecarboxamide, 97%
C5H7N3O
M75802
3-Methylpyrazole, 97%
C4H6N2