EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Substantiation of maximum permissible levels of mono-, di- and triisopropanolamine in the air of work areas].
PMID 3744114
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
110248
Amino-2-propanol, 93%
C3H9NO
W396508
DL-1-Amino-2-propanol, ≥90%
C3H9NO