EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Sepharose chromatography of transfer ribonucleic acids using reverse salt gradients.
PMID 374940
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09978
Ammonium sulfate, BioUltra, ≥99.0% (T)
H8N2O4S