EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Reconstitution of 50 S ribosomal subunits from Escherichia coli.
PMID 374950
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O