EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
The role of metaphosphate in the activation of the nucleotide by TPS and DCC in the oligonucleotide synthesis.
PMID 3754328
Abstract
The course of the activation of 3'-acetylthymidine 5'-phosphate by TPS and DCC were followed up by 31P FT nmr. The fact that the "metaphosphate" (delta-5.1) first becomes detectable only at later stage of the activation and does coexist with pyrophosphate and triphosphate suggests that the pyridinium derivative of "metaphosphate" is most probably not directly formed from the hypothetical mixed anhydride or active "pseudourea" right at the beginning of the reaction of pdTac with TPS or DCC, but rather formed at later stage of the activation reaction from the degradation of the pyro- and triphosphates by the activating agent. The mixed anhydride or the active "pseudourea" is most possibly the active key intermediate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119490
2,4,6-Triisopropylbenzenesulfonyl chloride, 97%
C15H23ClO2S