EMAIL THIS PAGE TO A FRIEND
Cancer letters
Formation of an N-nitrosamine by oxidation.
PMID 3756844
Abstract
Tolazamide, an antidiabetic drug, was found to produce N-nitrosohexamethyleneimine (NHM) upon exposure to an oxidizing agent and in the absence of a nitrosating agent. The oxidizing agents were either hydrogen peroxide or oxygen.