EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
More on fructosamine assay as index to glycated protein.
PMID 3757248
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6149
1-Deoxy-1-morpholino-D-fructose
C10H19NO6