EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Assessment of a potential dopaminergic prodrug moiety in several ring systems.
PMID 3761320
Abstract
The ortho hydroxy/methyl, hydroxy/hydroxymethyl, hydroxy/formyl, and hydroxy/carboxy substitution patterns, some of which confer dopaminergic agonist effects upon 2-aminotetralin ring systems, have been incorporated into beta-phenethylamine, 2-aminoindan, and trans-octahydrobenzo[f]quinoline rings. Certain of the 2-aminoindan derivatives displayed pharmacologic properties consistent with their being dopaminergic agonists. The beta-phenethylamine derivative did not show any significant dopamine-like activity. The 7-hydroxy-8-methyloctahydrobenzo[f]quinoline derivative 4a was a moderately potent, short-acting DA2 receptor antagonist. All of the carboxylic acid derivatives were inert. Of the ortho hydroxy/methyl derivatives, only the 5-hydroxy-6-methyl-2-aminotetralin derivative displayed pharmacological properties consistent with its being a dopaminergic prodrug. It is concluded that 5-hydroxy-6-methyl-2-(di-n-propylamino)tetralin (1a) is structurally unique for a dopaminergic drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
479128
2-Aminoindan, 97%
C9H11N
A59522
2-Aminoindan hydrochloride, 98%
C9H11N · HCl