EMAIL THIS PAGE TO A FRIEND
European journal of immunology
A human thymocyte antigen defined by a hybrid myeloma monoclonal antibody.
PMID 376318
Abstract
Spleen cells from a BALB/c mouse that had been immunized with human thymocytes were fused with the myeloma line P3-NS 1/1 Ag 4.1. One of the resulting hybrid clones (NA 1/34) secreted an antibody that was highly specific for human thymocytes. Eighty-five % of thymocytes expressed the antigen designated HTA1. There were an estimated 15 x 10(4) molecules of HTA 1 per cell, and it is therefore a major surface molecule. The expression of this antigen on thymocytes appears to be reciprocal to HLA, as recognized by another monoclonal antibody W6/32. Immunoprecipitated material from [125I]-labeled thymocyte membranes was analyzed by polyacrylamide gel electrophoresis in sodium dodecyl sulfate which disclosed a single component of 45,000 molecular weight.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
NA1/34.HLK, 85102243, Reactivity: Anti-CD1a, human