EMAIL THIS PAGE TO A FRIEND
Advances in biochemical psychopharmacology
GABA-mimetic compounds block haloperidol-induced hyperprolactinemia in rats.
PMID 3766216
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06720
2-Aminoethyl hydrogen sulfate, ≥98.0% (T)
C2H7NO4S