EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Metabolism and carcinogenicity of nitrotoluenes.
PMID 3766281
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N27314
3-Nitrotoluene, 99%
C7H7NO2
45986
3-Nitrotoluene, analytical standard
C7H7NO2