EMAIL THIS PAGE TO A FRIEND
British journal of clinical pharmacology
Ethoxyresorufin O-deethylation by human liver microsomes.
PMID 3768238
Abstract
As a substrate for human liver microsomes, ethoxyresorufin appears to be metabolised by a group of cytochrome P450 isoenzymes which are inducible by cigarette smoking. Kinetic studies in microsomes from four human livers indicate that only one enzyme component is involved over the full substrate range. Ethoxyresorufin O-deethylation activity at both high and low substrate concentrations correlated with the high affinity component of ethoxycoumarin O-deethylation and of diphenyloxazole metabolism.