EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Measurement of magnesium in serum and urine with a random access analyzer by use of a modified xylidyl blue-1 procedure.
PMID 3769208
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248266
Xylidyl blue I
C25H20N3NaO6S