EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Production of trans-o-hydroxybenzalpyruvate from 1,2-dihydroxynaphthalene by cell-free extracts of Corynebacterium renale.
PMID 3770791
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
232505
1,2-Dihydroxynaphthalene, technical grade
C10H8O2