EMAIL THIS PAGE TO A FRIEND
Przeglad lekarski
[Positive inotropic effect of digitalis on the left ventricular systolic time interval in patients with primary dilated cardiomyopathy observed during increased heart ventricle load].
PMID 3775049
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19