EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Formation in aqueous solution of N-nitroso curzate and the catalytic effect of some anions.
PMID 3779151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34326
Cymoxanil, PESTANAL®, analytical standard
C7H10N4O3