EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic regulation of the intermediate products of the production of ethsane--cyclohexane hydrochloride and ethylcyclohexylamine hydrochloride in various types of bodies of water].
PMID 3781284
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45408
Cycloate, PESTANAL®, analytical standard
C11H21NOS