EMAIL THIS PAGE TO A FRIEND
Medicine, science, and the law
Usefulness of gas chromatography/negative ion chemical ionization mass spectrometry for detection of an organophosphate pesticide in a victim.
PMID 3784847
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31611
Phenthoate, PESTANAL®, analytical standard
C12H17O4PS2