EMAIL THIS PAGE TO A FRIEND
Mutation research
Induction of chromosome aberrations by 90Sr beta-particles in cultured human lymphocytes.
PMID 3785263
Abstract
Human blood was irradiated with beta-particles from an external source of 90Sr. The source was a rolled piece of silver foil, active dimensions: 100 X 12.5 mm, incorporating 3.7 X 10(8) Bq (10 mCi) of 90Sr/90Y. After culturing for 48 h, the dicentric yield in the lymphocytes at the first metaphase was measured as a function of the dose in the blood. The aberration yield fitted the linear-quadratic function well, which is consistent with the single-track and two-track model for aberration formation at low LET radiation. The curve for 90Sr beta-rays was compared with a curve for 60Co gamma-rays. The main difference between the coefficients was in the alpha values. With respect to 60Co gamma-rays, the RBE calculated from the dose-effect relationships for dicentric production was 2.8 at the dose of 0.14 Gy; it decreased with increasing doses. The distribution of dicentrics was consistent with the Poisson distribution but showed a tendency to over-dispersion in the region of higher doses. A reason for these discrepancies is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF90975516
Silver, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95+%
Ag
GF23006797
Silver, foil, 25x25mm, thickness 0.1mm, annealed, 99.95+%
Ag
GF29990661
Silver, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95+%
Ag
GF74445813
Silver, foil, 25x25mm, thickness 0.20mm, as rolled, 99.95+%
Ag
GF70015946
Silver, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95+%
Ag
GF46518903
Silver, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Ag
GF10931947
Silver, foil, 0.5m coil, thickness 0.01mm, 99.95+%
Ag
GF64714215
Silver, foil, 0.5m coil, thickness 0.025mm, annealed, 99.95+%
Ag
GF40017620
Silver, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.95+%
Ag
GF62467386
Silver, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Ag
GF24663012
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Ag
GF51746250
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Ag
GF10306987
Silver, foil, 0.5m coil, thickness 0.05mm, annealed, 99.95+%
Ag
GF40779726
Silver, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Ag
GF83898905
Silver, foil, 0.5m coil, thickness 0.125mm, annealed, 99.95+%
Ag
GF71813785
Silver, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Ag
GF27190762
Silver, foil, 0.5m coil, thickness 0.125mm, coil width 300mm, annealed, 99.95+%
Ag
GF07805695
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Ag
GF23632332
Silver, foil, 0.5m coil, thickness 0.1mm, annealed, 99.95+%
Ag
GF58225518
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Ag
GF65202599
Silver, foil, 10mm disks, thickness 0.0025mm, 99.97%
Ag
GF96071104
Silver, foil, 10mm disks, thickness 0.003mm, 99.9%
Ag
GF78575732
Silver, foil, 10mm disks, thickness 0.004mm, 99.97%
Ag
GF05418312
Silver, foil, 10mm disks, thickness 0.005mm, 99.97%
Ag
GF35518169
Silver, foil, 10mm disks, thickness 0.007mm, 99.97%
Ag
GF25600777
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Ag
GF15732402
Silver, foil, 10mm disks, thickness 0.008mm, 99.97%
Ag
GF15118253
Silver, foil, 10mm disks, thickness 0.009mm, 99.97%
Ag
GF25755217
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Ag
GF92043244
Silver, foil, 10mm disks, thickness 0.015mm, 99.97%
Ag