EMAIL THIS PAGE TO A FRIEND
Toxicology and industrial health
Methylhistidine changes in tibialis anterior muscles of 6-mercaptopurine-treated rats.
PMID 3787652
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67520
1-Methyl-L-histidine, ≥98.0% (TLC)
C7H11N3O2