EMAIL THIS PAGE TO A FRIEND
Nihon eiseigaku zasshi. Japanese journal of hygiene
[The relation between the humoral immune response and the concentration of beryllium in blood and spleen of mice intraperitoneally injected with BeCl2].
PMID 3795588