EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Xanthine oxidase-catalyzed DNA binding of dihydrodiol derivatives of nitro-polycyclic aromatic hydrocarbons.
PMID 3800998
Abstract
Xanthine oxidase, a mammalian nitroreductase, catalyzed the covalent binding of a series of nitro-polycyclic aromatic hydrocarbons (nitro-PAHs) trans-dihydrodiols to DNA. Some of the trans-dihydrodiols bound to DNA to a greater extent than their parent nitro-PAHs; however, when the dihydrodiol moiety was peri to the nitro substituent low levels of binding were observed. These data illustrate that ring-oxidation and hydrolysis of nitro-PAHs to their trans-dihydrodiols followed by nitroreduction is a potential metabolic pathway leading to DNA adducts in mammals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10209
9-Nitroanthracene, 93%
C14H9NO2
BCR308
9-Nitroanthracene, BCR® certified Reference Material
C14H9NO2