EMAIL THIS PAGE TO A FRIEND
Clinical pharmacokinetics
Hypoalbuminaemia and naproxen pharmacokinetics in a patient with rheumatoid arthritis.
PMID 3802691
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC305
O-Desmethylnaproxen
C13H12O3