EMAIL THIS PAGE TO A FRIEND
Journal of Tongji Medical University = Tong ji yi ke da xue xue bao
[Effect of strophanthin, digoxin and isoproterenol on heart contractility in hypoxia and hypercapnic acidosis].
PMID 3806739
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30030
Cymarin, ≥96% (HPLC)
C30H44O9