EMAIL THIS PAGE TO A FRIEND
Molekuliarnaia biologiia
[Theoretical conformation analysis of beta-casomorphin-5 molecule].
PMID 3807911
Abstract
The spatial structure of beta-casomorphin-5 molecule--H-Tyr-Pro-Phe-Pro-Gly-OH has been investigated by the theoretical conformational analysis method. The results indicate that the conformational properties of the molecule can be represented by a large number of structures having twelve different backbone forms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5147
β-Casomorphin Fragment 1-5 hydrochloride, ≥97% (HPLC)
C30H37N5O7