EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Measurement of cytoplasmic, free magnesium concentration with entrapped eriochrome blue in nerve endings isolated from the guinea pig brain.
PMID 3808456
Abstract
Cytoplasmic, free Mg2+ was measured spectrophotometrically using an intrasynaptosomally entrapped Mg2+-indicator, Eriochrome blue (EB). Addition of the ionophore A23187 or disruption of the synaptosomal plasma membrane with digitonin caused an increase in absorbance of entrapped EB with a maximum at 551 nm, which is typical for the Mg2+-EB complex. A conversion of absorbance changes to levels of free Mg2+ concentrations was performed after disruption of synaptosomal plasma membranes by digitonin. The results indicated that the internal, free Mg2+ increased from 0.34 to 2.2 mM when the extracellular Mg2+ concentration was increased from 1 to 5 mM. The low values of cytoplasmic, free Mg2+ concentrations suggest the presence of effective regulatory mechanisms in the nerve endings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201723
Plasmocorinth B, Dye content 60 %
C16H9ClN2Na2O9S2