EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Cytogenetic and embryotoxic effects of bromophos and demethylbromophos.
PMID 3809618
Abstract
The organophosphorus pesticide bromophos and the tetramethylammonium and sodium salts of demethylbromophos were tested for cytogenetic and embryotoxic activity on mice of different strains. Single intraperitoneal (ip) doses of 183.0 mg/kg (0.5 mmol/kg) and 73.2 mg/kg (0.2 mmol/kg) bromophos caused a significant enhancement in the percentage of chromosome aberrations in bone marrow cells of CFLP mice; similar effects were produced by a single dose of 0.2 mmol/kg demethylbromophos tetramethylammonium salt and demethylbromophos sodium salt trihydrate, respectively, indicating that the cytogenetic activity of bromophos is not connected with its alkylating properties. After repeated ip or oral administration to pregnant mice of strains AB Jena/Halle and DBA, none of the tested compounds showed a marked influence on the total implantation losses, although in some cases the postimplantation losses were significantly increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33400
Bromophos-methyl, PESTANAL®, analytical standard
C8H8BrCl2O3PS