EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Piperazine toxicity in a kitten.
PMID 3811191
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46632
Citric acid sesquipiperazine salt hydrate, VETRANAL®, analytical standard
C6H8O7 · 1.5C4H10N2 · xH2O