EMAIL THIS PAGE TO A FRIEND
Akusherstvo i ginekologiia
[Suppression of unwanted lactation in parturients with estrogens and bromocriptine].
PMID 3812907
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3881
Diethylstilbestrol dipropionate, analytical standard, for drug analysis
C24H28O4