EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Capillary gas chromatography/pulsed supersonic jet/fluorescence excitation spectroscopy for the identification of methylanthracenes in a complex environmental sample.
PMID 3813017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M29401
2-Methylanthracene, 97%
C15H12
M29606
9-Methylanthracene, 98%
C15H12