EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of beryllium nitrate on early and late pregnancy in rats.
PMID 3814854
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14205
Beryllium standard for AAS, analytical standard, 1.0 g/L Be in nitric acid
BeN2O6