EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Regiospecific deoxygenation of the dihydroxyacetone moiety at C-17 of corticoid steroids with iodotrimethylsilane.
PMID 3815593
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195529
Iodotrimethylsilane, 97%
C3H9ISi