EMAIL THIS PAGE TO A FRIEND
Giornale italiano di chemioterapia
[In vitro antiviral activity of octophene].
PMID 3817378
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2290
Clofoctol, analytical standard
C21H26Cl2O