EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
[Effect of aluminum ion on sulfisomidine absorption in rabbits].
PMID 3820089
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46908
Sulfisomidin, VETRANAL®, analytical standard
C12H14N4O2S