EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
[Effect on the central nervous system and circulatory system of new derivatives of perhydroazepine, perhydro-1,5-oxazocine, morpholine and piperazine containing a pharmacophoric 3-(isopropylamino)-propan-2-ol group].
PMID 3834766
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H10401
Hexamethyleneimine, 99%
C6H13N