EMAIL THIS PAGE TO A FRIEND
Zhonghua liu xing bing xue za zhi = Zhonghua liuxingbingxue zazhi
[Single dose of 2 acute rodenticides used in a village].
PMID 3836748
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36564
Crimidine, PESTANAL®, analytical standard
C7H10ClN3