EMAIL THIS PAGE TO A FRIEND
Doklady Akademii nauk SSSR
[Chemiluminescence of peritoneal macrophages activated by non-natural polyelectrolytes].
PMID 3839743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2667
Sequa-brene, gelatinous solid