EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Palladium-catalyzed cross-coupling of 2-iodoadenosine with terminal alkynes: synthesis and biological activities of 2-alkynyladenosines.
PMID 3840058
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
663115
2-Iodoadenosine, 97%
C10H12IN5O4