EMAIL THIS PAGE TO A FRIEND
BioEssays : news and reviews in molecular, cellular and developmental biology
BUdR, probability and cell variants: towards a molecular understanding of the decision to differentiate.
PMID 3842598